CAS RN 2497-18-9
IUPAC Name:
(2Z)-hex-2-en-1-yl acetate
Synonyme:
trans-2-hexenyl acetate
Molekulargewicht (g/mol):
142.2
Summenformel:
C8H14O2
InChi Key:
HRHOWZHRCRZVCU-WAYWQWQTSA-N
SMILES:
CCC\C=C/COC(C)=O
1
–
1
von
1
Ergebnisse
1
trans-2-Hexenylacetat, 98 %, Thermo Scientific Chemicals
CAS: 2497-18-9 Summenformel: C8H14O2 Molekulargewicht (g/mol): 142.198 MDL-Nummer: MFCD00009474 InChI-Schlüssel: HRHOWZHRCRZVCU-AATRIKPKSA-N Synonym: z-hex-2-enyl acetate,unii-pt792bq57d,2-hexen-1-ol, acetate, z,z-2-hexenyl acetate,fema no. 2564,2-hexen-1-yl acetate, trans,2-hexen-1-ol, acetate, 2e,2-hexenyl acetate, trans,hexenyl acetate, 2z PubChem CID: 2733294 IUPAC-Name: [(E)-Hex-2-enyl]-acetat SMILES: CCCC=CCOC(=O)C