CAS RN 25100-12-3
IUPAC Name:
hydrogen N-cyclohexylhydroxylamine chloride
Synonyme:
hydrogen N-cyclohexylhydroxylamine chloride
Molekulargewicht (g/mol):
151.63
Summenformel:
C6H14ClNO
InChi Key:
SSVAHXZUFFSFER-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].ONC1CCCCC1
1
–
1
von
1
Ergebnisse
1
N-Cyclohexylhydroxylaminhydrochlorid, 98 %, Thermo Scientific Chemicals
CAS: 25100-12-3 Summenformel: C6H14ClNO Molekulargewicht (g/mol): 151.634 MDL-Nummer: MFCD00012565 InChI-Schlüssel: SSVAHXZUFFSFER-UHFFFAOYSA-N Synonym: n-cyclohexylhydroxylamine hydrochloride,cyclohexylhydroxylamine hydrochloride,unii-r98vsp3t5l,r98vsp3t5l,cyclohexylhydroxylamine hcl,acmc-1ch2g,n-cyclohexylhydroxylarnine hydrochloride,n-cyclohexyl hydroxylamine hydrochloride,cyclohexanamine,n-hydroxy-,hydrochloride 1:1 PubChem CID: 3084432 IUPAC-Name: N-Cyclohexylhydroxylamin;hydrochlorid SMILES: C1CCC(CC1)NO.Cl