CAS RN 2739-12-0
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
N-Methylaniline Hydrochloride, TRC
CAS: 2739-12-0 Summenformel: C7H9N . HCl Molekulargewicht (g/mol): 143.61 Synonym: N-Methylanilinium Chloride,N-Methylbenzenamine Hydrochloride,Anilinomethane Hydrochloride IUPAC-Name: N-methylaniline;hydrochloride SMILES: Cl.CNc1ccccc1