CAS RN 307496-26-0
CAS RN 307496-26-0
IUPAC Name:
(2,4-difluorophenyl)methanide; bromozincylium
Synonyme:
(2,4-difluorophenyl)methanide; bromozincylium
Molekulargewicht (g/mol):
272.4
Summenformel:
C7H5BrF2Zn
InChi Key:
WPJAHFYOACWPNA-UHFFFAOYSA-M
SMILES:
[Zn+]Br.[CH2-]C1=C(F)C=C(F)C=C1