CAS RN 33401-94-4
CAS RN 33401-94-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Pyrantel tartrate, MedChemExpress
MedChemExpress Pyrantel tartrate, a tetrahydropyrimidine broad-spectrum anthelmintic, and is a nicotinic acetylcholine receptor (nAChR) agonist. Pyrantel tartrate can elicit spastic muscle paralysis in parasitic worms. Pyrantel tartrate can be used for the research of astrointestinal nematodes infections.