CAS RN 33462-81-6
IUPAC Name:
cyclopropyldiphenylsulfanium; tetrafluoroboranuide
Synonyme:
cyclopropyldiphenylsulfanium tetrafluoroborate
Molekulargewicht (g/mol):
314.15
Summenformel:
C15H15BF4S
InChi Key:
DWEYKSWKFYZEGZ-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.C1CC1[S+](C1=CC=CC=C1)C1=CC=CC=C1