CAS RN 334949-59-6
CAS RN 334949-59-6
IUPAC Name:
3-[({3-[(dimethylamino)methyl]phenyl}imino)(phenyl)methyl]-2-hydroxy-1H-indole-6-carboxamide
Synonyme:
3-[({3-[(dimethylamino)methyl]phenyl}imino)(phenyl)methyl]-2-hydroxy-1H-indole-6-carboxamide
Molekulargewicht (g/mol):
412.49
Summenformel:
C25H24N4O2
InChi Key:
WGPXKFOFEXJMBD-UHFFFAOYSA-N
SMILES:
CN(C)CC1=CC(=CC=C1)N=C(C1=C(O)NC2=CC(=CC=C12)C(N)=O)C1=CC=CC=C1
1
–
2
von
2
Ergebnisse
2
BIX02188, MedChemExpress
MedChemExpress BIX02188 is a potent MEK5-selective inhibitor with an IC50 of 4.3 nM. BIX02188 inhibits ERK5 catalytic activity, with an IC50 of 810 nM.