CAS RN 335-48-8
IUPAC Name:
1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane
Synonyme:
1,4-dibromooctafluorobutane
Molekulargewicht (g/mol):
359.84
Summenformel:
C4Br2F8
InChi Key:
RWWUGYJWSVESJC-UHFFFAOYSA-N
SMILES:
FC(F)(Br)C(F)(F)C(F)(F)C(F)(F)Br
1
–
1
von
1
Ergebnisse
1
1,4-Dibromoctafluorbutan, 98 %, Thermo Scientific™
CAS: 335-48-8 Summenformel: C4Br2F8 Molekulargewicht (g/mol): 359.839 MDL-Nummer: MFCD00153112 InChI-Schlüssel: RWWUGYJWSVESJC-UHFFFAOYSA-N Synonym: 1,4-dibromooctafluorobutane,1,4-dibromoperfluorobutane,butane, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro,acmc-209i1o,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro-butane,butane,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro,1,4-bis bromanyl-1,1,2,2,3,3,4,4-octakis fluoranyl butane PubChem CID: 2736783 IUPAC-Name: 1,4-Dibrom-1,1,2,2,3,3,4,4-Octafluorbutan SMILES: C(C(C(F)(F)Br)(F)F)(C(F)(F)Br)(F)F