CAS RN 33693-04-8
IUPAC Name:
N2-tert-butyl-N4-ethyl-6-methoxy-1,3,5-triazine-2,4-diamine
Synonyme:
terbumeton
Molekulargewicht (g/mol):
225.3
Summenformel:
C10H19N5O
InChi Key:
BCQMBFHBDZVHKU-UHFFFAOYSA-N
SMILES:
CCNC1=NC(NC(C)(C)C)=NC(OC)=N1