CAS RN 3848-24-6
CAS RN 3848-24-6
IUPAC Name:
hexane-2,3-dione
Synonyme:
2,3-hexanedione
Molekulargewicht (g/mol):
114.14
Summenformel:
C6H10O2
InChi Key:
MWVFCEVNXHTDNF-UHFFFAOYSA-N
SMILES:
CCCC(=O)C(C)=O
1
–
1
von
1
Ergebnisse
1
2,3-Hexandion, 94 %, Thermo Scientific Chemicals
CAS: 3848-24-6 Summenformel: C6H10O2 Molekulargewicht (g/mol): 114.14 MDL-Nummer: MFCD00009398 InChI-Schlüssel: MWVFCEVNXHTDNF-UHFFFAOYSA-N Synonym: 2,3-hexanedione,acetylbutyryl,acetyl butyryl,methyl propyl diketone,butyryl acetyl,hexanedione,unii-559anr3nvs,2,3-hexandione,fema no. 2558,559anr3nvs PubChem CID: 19707 ChEBI: CHEBI:87583 IUPAC-Name: Hexan-2,3-Dion SMILES: CCCC(=O)C(C)=O