CAS RN 38771-21-0
CAS RN 38771-21-0
IUPAC Name:
4-bromobut-1-yne
Synonyme:
4-bromobut-1-yne
Molekulargewicht (g/mol):
132.99
Summenformel:
C4H5Br
InChi Key:
XLYOGWXIKVUXCL-UHFFFAOYSA-N
SMILES:
BrCCC#C
1
–
2
von
2
Ergebnisse
1
4-Bromo-1-Butin, 97 %, Thermo Scientific Chemicals
CAS: 38771-21-0 Summenformel: C4H5Br Molekulargewicht (g/mol): 132.988 MDL-Nummer: MFCD10000883 InChI-Schlüssel: XLYOGWXIKVUXCL-UHFFFAOYSA-N Synonym: 4-bromo-1-butyne,1-bromo-3-butyne,1-butyne, 4-bromo,4-bromobutyne,3-butynyl bromide,1-bromo-3-butynol,4-bromo-but-1-yne,acmc-1afup,1-butyne, 4-bromo-6ci,9ci PubChem CID: 11073464 IUPAC-Name: 4-Brombut-1-yn SMILES: C#CCCBr
2
4-Bromo-1-butyne, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.