CAS RN 39513-75-2
IUPAC Name:
6-methyl-3,4-dihydro-2H-1-benzopyran-4-one
Synonyme:
6-methyl-2,3-dihydro-1-benzopyran-4-one
Molekulargewicht (g/mol):
162.19
Summenformel:
C10H10O2
InChi Key:
RJHXEPLSJAVTFW-UHFFFAOYSA-N
SMILES:
CC1=CC2=C(OCCC2=O)C=C1