CAS RN 50593-92-5
IUPAC Name:
5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylic acid
Synonyme:
5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylic acid
Molekulargewicht (g/mol):
249.08
Summenformel:
C6H5BrN2O2S
InChi Key:
YJEWVVYJOJJLBP-UHFFFAOYSA-N
SMILES:
CSC1=NC=C(Br)C(=N1)C(O)=O