CAS RN 51011-05-3
CAS RN 51011-05-3
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
MedChemExpress 6″-O-Malonylgenistin
MedChemExpress 6″-O-Malonylgenistin(Malonylgenistin) is an isoflavone derivative.