CAS RN 516-03-0
CAS RN 516-03-0
IUPAC Name:
λ²-iron(2+) oxalate
Synonyme:
λ²-iron(2+) oxalate
Molekulargewicht (g/mol):
143.86
Summenformel:
C2FeO4
InChi Key:
OWZIYWAUNZMLRT-UHFFFAOYSA-L
SMILES:
[Fe++].[O-]C(=O)C([O-])=O
1
–
1
von
1
Ergebnisse
1
Eisen(II)-Oxalathydrathydrat, 96 %, Thermo Scientific Chemicals
CAS: 516-03-0 Summenformel: C2FeO4 Molekulargewicht (g/mol): 143.86 MDL-Nummer: MFCD00012475 InChI-Schlüssel: OWZIYWAUNZMLRT-UHFFFAOYSA-L Synonym: ferrous oxalate,iron ii oxalate,unii-dzp4yv3icv,dzp4yv3icv,iron protoxalate,iron, ethanedioato 2--.kappa.o1,.kappa.o2,iron 2+ oxalate,iron oxalate fec2o4,hsdb 463,iron 2+ ; oxalate PubChem CID: 10589 IUPAC-Name: λ²-iron(2+) oxalate SMILES: [Fe++].[O-]C(=O)C([O-])=O