CAS RN 536-60-7
CAS RN 536-60-7
IUPAC Name:
[4-(propan-2-yl)phenyl]methanol
Synonyme:
cuminol cuminyl alcohol cuminic alcohol cumin alcohol
Molekulargewicht (g/mol):
150.22
Summenformel:
C10H14O
InChi Key:
OIGWAXDAPKFNCQ-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=C(CO)C=C1
1
–
1
von
1
Ergebnisse
1
4-Isopropylbenzylalkohol, 97 %, Thermo Scientific Chemicals
CAS: 536-60-7 Summenformel: C10H14O Molekulargewicht (g/mol): 150.22 MDL-Nummer: MFCD00004663 InChI-Schlüssel: OIGWAXDAPKFNCQ-UHFFFAOYSA-N Synonym: 4-isopropylbenzyl alcohol,cuminol,cumic alcohol,cuminyl alcohol,cuminic alcohol,4-isopropylphenyl methanol,cumyl alcohol,p-cymen-7-ol,cumin alcohol,p-isopropylbenzyl alcohol PubChem CID: 325 ChEBI: CHEBI:27628 IUPAC-Name: (4-propan -2-ylphenyl)methanol SMILES: CC(C)C1=CC=C(C=C1)CO