CAS RN 54375-47-2
CAS RN 54375-47-2
IUPAC Name:
2-[(carboxylatomethyl)[(7-hydroxy-4-methyl-2-oxo-2H-chromen-8-yl)methyl]azaniumyl]acetate
Synonyme:
2-[(carboxylatomethyl)[(7-hydroxy-4-methyl-2-oxochromen-8-yl)methyl]ammonio]acetate
Molekulargewicht (g/mol):
320.28
Summenformel:
C15H14NO7
InChi Key:
RJPSHDMGSVVHFA-UHFFFAOYSA-M
SMILES:
CC1=CC(=O)OC2=C(C[NH+](CC([O-])=O)CC([O-])=O)C(O)=CC=C12
1
–
1
von
1
Ergebnisse
1
MedChemExpress Calcein Blue
MedChemExpress Calcein Blue, a membrane-impermeant fluorescent dye, is a coumarin derivative that contains an iminodiacetic acid structure. Calcein Blue is also a metallofluorochromic indicator.