CAS RN 557-28-8
CAS RN 557-28-8
IUPAC Name:
zinc(2+) dipropanoate
Synonyme:
zinc(2+) dipropionate
Molekulargewicht (g/mol):
211.52
Summenformel:
C6H10O4Zn
InChi Key:
XDWXRAYGALQIFG-UHFFFAOYSA-L
SMILES:
[Zn++].CCC([O-])=O.CCC([O-])=O
1
–
1
von
1
Ergebnisse
1
Zinkpropionat, 97 %, Thermo Scientific Chemicals
CAS: 557-28-8 Summenformel: C6H10O4Zn Molekulargewicht (g/mol): 211.52 MDL-Nummer: MFCD00045937 InChI-Schlüssel: XDWXRAYGALQIFG-UHFFFAOYSA-L Synonym: zinc propionate,unii-81pxa87xvc,81pxa87xvc,propanoic acid, zinc salt 2:1,gyna-plex,propionic acid, zinc salt,zinc dipropionate,propanoic acid, zinc salt,zinc propanoate,propanoic acid, zinc salt, basic PubChem CID: 11189 IUPAC-Name: Zink;propanoat SMILES: [Zn++].CCC([O-])=O.CCC([O-])=O