CAS RN 589-68-4
IUPAC Name:
2,3-dihydroxypropyl tetradecanoate
Synonyme:
2,3-dihydroxypropyl tetradecanoate
Molekulargewicht (g/mol):
302.46
Summenformel:
C17H34O4
InChi Key:
DCBSHORRWZKAKO-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCCCC(=O)OCC(O)CO