CAS RN 6223-35-4
CAS RN 6223-35-4
IUPAC Name:
sodium 3,8-dimethyl-5-(propan-2-yl)azulene-1-sulfonate
Synonyme:
sodium guaiazulene sulfonate
Molekulargewicht (g/mol):
300.35
Summenformel:
C15H17NaO3S
InChi Key:
GEYJUFBPCGDENK-UHFFFAOYSA-M
SMILES:
[Na+].CC(C)C1=CC=C(C)C2=C(C=C(C)C2=C1)S([O-])(=O)=O
1
–
1
von
1
Ergebnisse
1
Sodium gualenate, MedChemExpress
MedChemExpress Sodium gualenate (Guaiazulenesulfonate sodium) is a hydrophilic derivative of guaiazulene with excellent anti-inflammatory and wound-healing effects mainly used for the treatment of duodenal ulcer, gastric ulcer and gastritis.