CAS RN 65358-15-8
CAS RN 65358-15-8
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
6
von
6
Ergebnisse
1
MedChemExpress Pseudothymidine
MedChemExpress Pseudothymidine is a C-nucleoside analog of thymidine.
2
MedChemExpress Pseudothymidine
MedChemExpress Pseudothymidine is a C-nucleoside analog of thymidine.
3
MedChemExpress Pseudothymidine
MedChemExpress Pseudothymidine is a C-nucleoside analog of thymidine.
4
MedChemExpress Pseudothymidine
MedChemExpress Pseudothymidine is a C-nucleoside analog of thymidine.
5
MedChemExpress Pseudothymidine
MedChemExpress Pseudothymidine is a C-nucleoside analog of thymidine.
6
MedChemExpress Pseudothymidine
MedChemExpress Pseudothymidine is a C-nucleoside analog of thymidine.