CAS RN 68424-04-4
CAS RN 68424-04-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
4
von
4
Ergebnisse
1
Polydextrose, Thermo Scientific Chemicals
CAS: 68424-04-4 Summenformel: (C6H10O5)n MDL-Nummer: MFCD00804499 IUPAC-Name: Polydextrose SMILES: OCC1OC(-*)[C@@H](O)C(O)[C@@H]1O-*
2
Polydextrose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Polydextrose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Polydextrose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.