CAS RN 70-47-3
CAS RN 70-47-3
Thermo Scientific Chemicals L-(+)-Asparagin, 99 %
CAS: 70-47-3 Summenformel: C4H8N2O3 Molekulargewicht (g/mol): 132.119 MDL-Nummer: MFCD00064401 InChI-Schlüssel: DCXYFEDJOCDNAF-REOHCLBHSA-N Synonym: l-asparagine,asparagine,s-asparagine,aspartamic acid,agedoite,altheine,h-asn-oh,crystal vi,asparamide,asparagine acid PubChem CID: 6267 ChEBI: CHEBI:17196 IUPAC-Name: (2S)-2,4-diamino-4-Oxobutansäure SMILES: C(C(C(=O)O)N)C(=O)N
L-Asparagin, 99 %, Thermo Scientific Chemicals
CAS: 70-47-3 Summenformel: C4H8N2O3 Molekulargewicht (g/mol): 132.119 MDL-Nummer: MFCD00064401 InChI-Schlüssel: DCXYFEDJOCDNAF-REOHCLBHSA-N Synonym: l-asparagine,asparagine,s-asparagine,aspartamic acid,agedoite,altheine,h-asn-oh,crystal vi,asparamide,asparagine acid PubChem CID: 6267 ChEBI: CHEBI:17196 IUPAC-Name: (2S)-2,4-diamino-4-Oxobutansäure SMILES: C(C(C(=O)O)N)C(=O)N
L-Asparagin, wasserfrei,99 % [wasserfrei], MP Biomedicals
CAS: 70-47-3 Summenformel: C4H8N2O3 Molekulargewicht (g/mol): 132.119 InChI-Schlüssel: DCXYFEDJOCDNAF-REOHCLBHSA-N Synonym: L-Asparagin,Asparagin,s-Asparagin,Aspartaminsäure,Agedoit,Althein,Asparamid,Asparaginsäure,asparamide,asparagine acid PubChem CID: 6267 ChEBI: CHEBI:17196 IUPAC-Name: (2S)-2,4-diamino-4-Oxobutansäure SMILES: C(C(C(=O)O)N)C(=O)N
Melford L-Asparagin wasserfrei
Wasserfreies L-Asparagin wird häufig als Bestandteil von Zellkulturmedien verwendet.
L-Asparagine, TRC
CAS: 70-47-3 Summenformel: C4 H8 N2 O3 Molekulargewicht (g/mol): 132.12 Synonym: L-Asparagine,Asparagine, L- (8CI),(-)-Asparagine,(S)-2,4-Diamino-4-oxobutanoic acid,(S)-Asparagine,Agedoite,Altheine,Asn,Asparagine,Asparagine acid,Asparamide,Aspartamic acid,Aspartic acid amide,Aspartic acid β-amide,Butanoic acid, 2,4-diamino-4-oxo-, (S)-,Crystal VI,L-2,4-Diamino-4-oxobutanoic acid,L-Aspartamine,L-β-Asparagine,NSC 82391,l-Asparagine,α-Aminosuccinamic acid IUPAC-Name: (2S)-2,4-diamino-4-oxobutanoic acid SMILES: N[C@@H](CC(=O)N)C(=O)O
L-Asparagine, MedChemExpress
MedChemExpress L-Asparagine ((-)-Asparagine) is a non-essential amino acid that is involved in the metabolic control of cell functions in nerve and brain tissue.
L-Asparagin, wasserfrei, ≥ 98.5 %, Ultrapure, Thermo Scientific Chemicals
CAS: 70-47-3 Summenformel: C4H8N2O3 Molekulargewicht (g/mol): 132.119 MDL-Nummer: MFCD00064401 InChI-Schlüssel: DCXYFEDJOCDNAF-REOHCLBHSA-N Synonym: l-asparagine,asparagine,s-asparagine,aspartamic acid,agedoite,altheine,h-asn-oh,crystal vi,asparamide,asparagine acid PubChem CID: 6267 ChEBI: CHEBI:17196 IUPAC-Name: (2 S)-2,4-Diamino-4-Oxobutansäure SMILES: C(C(C(=O)O)N)C(=O)N