CAS RN 7439-95-4
Magnesium, ≥99.9 %, (Spurenmetallbasis), Drehspäne, Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesiumpulver, -325 Mesh, 99.8 %, Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesiumband, ungefähre Abmessungen: 3.0 mm (0.12 in.) Breite, 0.15 mm (0.0059 in.) Dicke, Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesium-Drehspäne, 99.8 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesium, Drehspäne, 99+ %, Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesium, Plasma-Standardlösung, Thermo Scientific Chemicals, Specpure™
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesiumgranulat, -4+30 mesh, 99.8 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesium-Drehspäne, -4 Mesh, 99.98 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesiumfolie, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesiumfolie, 1.0 mm (0.04 in.) Dicke, hart, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesium, AAS-Standardlösung, Specpure™, Mg 1000 μg/ml, Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]
Magnesiumkugel, 3.175 mm (0.125 Zoll) Durchmesser x 6.35 mm (0.25 Zoll) Länge, geglüht, 99.95 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7439-95-4 Summenformel: Mg Molekulargewicht (g/mol): 24.30 MDL-Nummer: MFCD00085308 InChI-Schlüssel: JLVVSXFLKOJNIY-UHFFFAOYSA-N Synonym: magnesio,sheet,powdered,magnesio italian,turnings,rieke's active,ribbon,unii-i38zp9992a,hsdb 654,nitrate, acs PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC-Name: Magnesium SMILES: [Mg]