CAS RN 7440-22-4
Silberpulver, -100 Mesh, 99.95 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberdraht, 2.0 mm (0.08 Zoll), hart, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberdraht, 0.1 mm (0.004 in.) Durchmesser, Premion™, 99.997 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberfolie, 0.075 mm (0.003 Zoll) Dicke, hart, 99.95 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberpulver, 4 bis 7 Mikrometer, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silber-Shot, 1 bis 7 mm (0.04 bis 0.3 Zoll), Premion™, 99.999 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberdraht, 0.25 mm (0.01 Zoll) im Durchmesser, hart, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberfolie, 1.0 mm (0.04 Zoll) Dicke, Premion™, 99.9985 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silber-Nanopartikel, 10 nm, 0.02 mg/ml, geliefert in 2 mM Natriumcitrat, 390 bis 400 nm-Absorption, Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberkugel, 3.175 mm (0.125 Zoll) Durchmesser x 6.35 mm (0.25 Zoll) Länge, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberkugel, 1–5 mm (0.04–0.2 in.), Premion™, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberdraht, 0.25 mm (0.01 Zoll) im Durchmesser, geglüht, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silber-Shot, 99.999 % (Metallbasis), Thermo Scientific Chemicals, Premion™
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]
Silberschläuche, 6.35 mm (0.25 Zoll) Außendurchmesser, 4.35 mm (0.171 Zoll) Innendurchmesser, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-22-4 Summenformel: Ag Molekulargewicht (g/mol): 107.87 MDL-Nummer: MFCD00003397 InChI-Schlüssel: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC-Name: Silber SMILES: [Ag]