CAS RN 7440-74-6
Indiumfolie, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumfolie, 0.25 mm Dicke, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumfolie, 99.9975 % (Metallbasis), Thermo Scientific Chemicals, Puratronic™
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumstücke, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumpulver, -100 Mesh, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumlösung für ICP, 1 g/l, HNO3 Matrix, Fisher Chemical™
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indium-Shot, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumdraht, 99.99 % (Metallbasis), Thermo Scientific Chemicals, Puratronic™
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumdraht, 1.0 mm (0.04 Zoll) Durchmesser, geglüht, Puratronic™, ≥99.998 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumpulver, -100 Mesh, 99.9 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumpulver, 99.99 % (Metallbasis), Thermo Scientific Chemicals, Puratronic™
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumkugeln, 1 mm (0.04 Zoll) Durchmesser, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumstab, 2.38 mm (0.094 Zoll) Durchmesser, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]
Indiumdraht, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-74-6 Summenformel: In Molekulargewicht (g/mol): 114.82 MDL-Nummer: MFCD00134048 InChI-Schlüssel: APFVFJFRJDLVQX-UHFFFAOYSA-N Synonym: indium, elemental,unii-045a6v3vfx,powder,shot,powder,-100 mesh,indio,shot, tear drop, 4mm 0.2in,ingot, polycrystalline, puratronic,shot, 5mm 0.2in & down, puratronic,wire, 0.5mm 0.02in dia, puratronic PubChem CID: 5359967 ChEBI: CHEBI:30430 IUPAC-Name: Indium SMILES: [In]