CAS RN 7798-23-4
CAS RN 7798-23-4
IUPAC Name:
tricopper(2+) diphosphate
Synonyme:
copper(II) phosphate
Molekulargewicht (g/mol):
380.58
Summenformel:
Cu3O8P2
InChi Key:
GQDHEYWVLBJKBA-UHFFFAOYSA-H
SMILES:
[Cu++].[Cu++].[Cu++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
1
–
1
von
1
Ergebnisse
1
Kupfer(II)-phosphat, 98 %, Thermo Scientific Chemicals
CAS: 7798-23-4 Summenformel: Cu3O8P2 Molekulargewicht (g/mol): 380.578 MDL-Nummer: MFCD00036268 InChI-Schlüssel: GQDHEYWVLBJKBA-UHFFFAOYSA-H Synonym: copper ii phosphate,cupric phosphate,unii-n8np6fr80r,tricopper bis orthophosphate,n8np6fr80r,copper phosphate cu3 po4 2,copper phosphate 3:2,hsdb 267,cupric phosphate cu3 po4 2,phosphoric acid, copper 2+ salt PubChem CID: 86469 IUPAC-Name: Kupfer(3+);diphosphat SMILES: [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cu+2].[Cu+2].[Cu+2]