CAS RN 9001-63-2
CAS RN 9001-63-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
MedChemExpress Lysozyme
MedChemExpress Lysozyme is an antimicrobial enzyme produced by animals that forms part of the innate immune system.
2
MedChemExpress Lysozyme
MedChemExpress Lysozyme is an antimicrobial enzyme produced by animals that forms part of the innate immune system.