CAS RN 9001-92-7
CAS RN 9001-92-7
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
MedChemExpress Proteinase
MedChemExpress Proteinase refers to the enzymes with proteolytic activity.