CAS RN 999-21-3
CAS RN 999-21-3
IUPAC Name:
1,4-bis(prop-2-en-1-yl) (2Z)-but-2-enedioate
Synonyme:
diallyl maleate
Molekulargewicht (g/mol):
196.2
Summenformel:
C10H12O4
InChi Key:
ZPOLOEWJWXZUSP-WAYWQWQTSA-N
SMILES:
C=CCOC(=O)\C=C/C(=O)OCC=C
1
–
1
von
1
Ergebnisse
1
Diallylmaleat, 95%, Thermo Scientific Chemicals
CAS: 999-21-3 Summenformel: C10H12O4 Molekulargewicht (g/mol): 196.202 MDL-Nummer: MFCD00010763 InChI-Schlüssel: ZPOLOEWJWXZUSP-WAYWQWQTSA-N Synonym: diallyl fumarate,fumaric acid, diallyl ester,unii-k852lk4c5p,2-butenedioic acid e-, di-2-propenyl ester,2-butenedioic acid 2e-, di-2-propenyl ester,butenedioic acid diallyl ester,1,4-bis prop-2-en-1-yl 2e-but-2-enedioate,2-butenedioic acid 2e-, 1,4-di-2-propen-1-yl ester,diallyl malleate,1,4-bis prop-2-en-1-yl but-2-enedioate PubChem CID: 5354266 IUPAC-Name: Bis(prop-2-enyl) (Z)-but-2-enedioat SMILES: C=CCOC(=O)C=CC(=O)OCC=C