missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dibrompinacolon, 98 %, Thermo Scientific™

Catalog Number 11467837
Click to view available options
Menge:
10 g
50 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
30263-65-1
C6H10Br2O
257.95
MFCD00045030
WIPDATICDHNWKG-UHFFFAOYSA-N
1,1-dibromopinacolone, dibromopinacolin, 2-butanone, 1,1-dibromo-3,3-dimethyl, omega,omega-dibrompinakolin german, omega,omega-dibrompinakolin, 4,4-dibromopinacolin, 3-01-00-02843 beilstein handbook reference
34866
1,1-dibromo-3,3-dimethylbutan-2-one
CC(C)(C)C(=O)C(Br)Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
30263-65-1
C6H10Br2O
257.95
MFCD00045030
WIPDATICDHNWKG-UHFFFAOYSA-N
1,1-dibromopinacolone, dibromopinacolin, 2-butanone, 1,1-dibromo-3,3-dimethyl, omega,omega-dibrompinakolin german, omega,omega-dibrompinakolin, 4,4-dibromopinacolin, 3-01-00-02843 beilstein handbook reference
34866
1,1-dibromo-3,3-dimethylbutan-2-one
CC(C)(C)C(=O)C(Br)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1,1-Dibromopinacolone
Siedepunkt 96°C to 97°C (12 mmHg)
Strukturformel (CH3)3CCOCHBr2
Menge 10 g
Formelmasse 257.95
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'