missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triazinane-2,4,6-trione, 98%, BLD Pharm™

Artikelnummer. 31030125
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
31030125 1 kg
31021895 500 g
31015449 100 g
31028452 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31030125 Lieferant BLD Pharm Lieferanten-Nr. BD1225341000g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Zum Warenkorb hinzufügen
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 1,3,5-Triazinane-2,4,6-trione
CAS 108-80-5
Summenformel C3H3N3O3
MDL-Nummer MFCD00082990
Menge 1 kg
SMILES O=C(NC(N1)=O)NC1=O
Molekulargewicht (g/mol) 129.07
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.