missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropyl-1'-(2-methyl-1H-benzo[d]imidazole-6-carbonyl)-4,6-dihydrospiro[indazole-5,4'-piperidin]-7(1H)-one, 99%, BLD Pharm™

Product Code. 31091617
Encompass_Preferred
Change view
Click to view available options
Menge:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31091617 1 mg
31083578 25 mg
31084914 50 mg
31077917 10 mg
31079214 5 mg
31090785 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31091617 Supplier BLD Pharm Supplier No. BD008311921mg
 Eingeschränkter Artikel
Verfügbar ab: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material 1-Isopropyl-1'-(2-methyl-1H-benzo[d]imidazole-6-carbonyl)-4,6-dihydrospiro[indazole-5,4'-piperidin]-7(1H)-one
CAS 1301214-47-0
Summenformel C23H27N5O2
MDL-Nummer MFCD28502250
Menge 1 mg
SMILES O=C1CC2(CCN(C(C3=CC=C4C(NC(C)=N4)=C3)=O)CC2)CC5=C1N(C(C)C)N=C5
Molekulargewicht (g/mol) 405.49
Reinheit (%) 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.