missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-propylimidazolium Tetrafluoroborate, 98%, BLD Pharm™

Artikelnummer. 30936493
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30936493 25 g
30935804 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30936493 Lieferant BLD Pharm Lieferanten-Nr. BD0081272025g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

98% HPLC,N

Specifications

Chemischer Name oder Material 1-Methyl-3-propylimidazolium Tetrafluoroborate
CAS 244193-48-4
Summenformel C7H13BF4N2
MDL-Nummer MFCD14156016
Menge 25 g
SMILES CCCN1C=[N+](C)C=C1.F[B-](F)(F)F
Molekulargewicht (g/mol) 212.00
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.