Learn More
1-n-Butyl-4-methylpyridinium Hexafluorphosphat, 99 %, Thermo Scientific Chemicals
Catalog Number 11317119
Click to view available options
Menge:
5 g
50 g
401788-99-6
C10H16F6NP
295.209
MFCD03095438
XBSZSSGZQFAUPF-UHFFFAOYSA-N
1-butyl-4-methylpyridinium hexafluorophosphate, 4mbppf6, 1-butyl-4-methylpyridin-1-ium hexafluorophosphate, acmc-209jbi, dsstox_cid_29152, dsstox_rid_83371, dsstox_gsid_49296
11254899
1-Butyl-4-Methylpyridin-1-ium;Hexafluorphosphat
CCCC[N+]1=CC=C(C=C1)C.F[P-](F)(F)(F)(F)F
401788-99-6
C10H16F6NP
295.209
MFCD03095438
XBSZSSGZQFAUPF-UHFFFAOYSA-N
1-butyl-4-methylpyridinium hexafluorophosphate, 4mbppf6, 1-butyl-4-methylpyridin-1-ium hexafluorophosphate, acmc-209jbi, dsstox_cid_29152, dsstox_rid_83371, dsstox_gsid_49296
11254899
1-Butyl-4-Methylpyridin-1-ium;Hexafluorphosphat
CCCC[N+]1=CC=C(C=C1)C.F[P-](F)(F)(F)(F)F
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1-n-Butyl-4-methylpyridinium hexafluorophosphate
Schmelzpunkt 45°C
Menge 5 g
Formelmasse 295.22
Reinheit (%) 99%