missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11-Maleimidundecansäure, Thermo Scientific Chemicals
Click to view available options
Menge:
5 g
25 g
100 g
Beschreibung
The maleimide functional group can be used to conjugate a variety of biomolecules such as enzymes and DNA to the polymer chain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Chemischer Name oder Material | 11-Maleimidoundecanoic acid |
Schmelzpunkt | 89°C to 90°C |
Farbe | Weiß |
Menge | 5 g |
Löslichkeitsinformationen | Soluble in methanol,and chloroform. |
Formelmasse | 281.35 |
Physikalische Form | Pulver |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'