Learn More
Ibidi 12 Well Chamber, Removable
Artikelnummer. 17266223
Encompass_Preferred
Ansicht ändern
Click to view available options
Typ:
Chambered Coverglass
Packungsgröße:
15 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Typ unitSize
17266223 Chambered Coverglass 15 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17266223 Lieferant Ibidi Lieferanten-Nr. 81201
Please to purchase this item. Need a web account? Register with us today!
12 Well Chamber, Removable is a chamber slide with a removable 12 well silicone chamber.
  • Chamber slide with a removable 12 well silicone chamber for cell culture and immunofluorescence staining
  • for use with upright and inverted microscopes
  • bottom material: microscopy glass
  • sterilized
Spezifikation
Typ Chambered Coverglass