Learn More
14-3-3 Sigma Mouse anti-Human, CoraLite 594, Clone: 3F10D9, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-66251-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
14-3-3 Sigma is a member 14-3-3 family of highly conserved proteins participating in diverse cellular processes including cell cycle progression. It is exclusively present in various epithelial cells, and alternatively named as human epithelial marker (HEM), or stratifin. Alteration of its expression has been linked to cancer.
Spezifikation
14-3-3 Sigma | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
IgG1 |
Immunofluorescence, Flow Cytometry | |
3F10D9 | |
CoraLite 594 | |
100 μL | |
2810 | |