Learn More
ACROBiosystems Human Neuropilin-1 / NRP1 / CD304 Protein, His Tag
Marke: ACROBiosystems NR1-H5228-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Neuropilin-1 (NRP1) is also known as Vascular endothelial cell growth factor 165 receptor (VEGF165R), CD antigen CD304, which belongs to the neuropilin family. The membrane-bound isoform 1 is a receptor involved in the development of the cardiovascular system, in angiogenesis, in the formation of certain neuronal circuits and in organogenesis outside the nervous system. It mediates the chemorepulsant activity of semaphorins. It binds to semaphorin 3A, The PLGF-2 isoform of PGF, The VEGF-165 isoform of VEGF and VEGF-B. Coexpression with KDR results in increased VEGF-165 binding to KDR as well as increased chemotaxis. It may regulate VEGF-induced angiogenesis. The soluble isoform 2 binds VEGF-165 and appears to inhibit its binding to cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.8 kDa. The protein migrates as 75-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH07533 | |
50 μg | |
Phe 22 - Lys 644 | |
NRP1,Neuropilin-1,NRP,VEGF165R,CD304 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
70.8 kDa | |
Human Neuropilin-1, His Tag (NR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession # AAH07533). | |
-20°C | |
Neuropilin-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |