Learn More
ACROBiosystems Human Cadherin-6 / KCAD / CDH6 Protein, His Tag
Marke: ACROBiosystems CA6-H5229-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Cadherin-6 (CDH6) is also known as Kidney cadherin (K-cadherin or KCAD), is a a type II classical cadherin from the cadherin superfamily. Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. CDH6 / KCAD contains five cadherin domains. CDH6 is highly expressed in brain, cerebellum, and kidney. Lung, pancreas, and gastric mucosa show a weak expression and also expressed in certain liver and kidney carcinomas.
This protein carries a polyhistidine tag at the C-terminus. The Human Cadherin-6 will be further processed into mature form (Ser 54-Ala 615). The protein has a calculated MW of 67.2 kDa (pro-form) and 63.4 kDa (mature-form). The protein migrates as 86-96Spezifikation
AAH00019 | |
100 μg | |
Thr 19 - Ala 615 | |
CDH6,CAD6,KCAD,K-cadherin,Cadherin-6 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |
67.2 kDa and 63.4 kDa | |
Human Cadherin-6, His Tag (CA6-H5229) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Ala 615 (Accession # AAH00019). | |
-20°C | |
Cadherin-6 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |