Learn More
ACROBiosystems Human IL-12A / NKSF1 / p35 Protein, Fc Tag
Marke: ACROBiosystems NK1-H5252-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Interleukin-12 (IL12) is also known as natural killer cell stimulatory factor (NKSF), cytotoxic lymphocyte maturation factor (CLMF) , is a heterodimeric cytokine encoded by two separate genes, IL-12A (p35) and IL-12B (p40). IL12 is naturally produced by dendritic cells, macrophages and human B-lymphoblastoid cells (NC-37) in response to antigenic stimulation. IL-12 is involved in the differentiation of naive T cells into Th0 cells and plays an important role in the activities of natural killer cells and T lymphocytes.IL-12 also has anti-angiogenic activity, which means it can block the formation of new blood vessels. Interleukin-12 subunit alpha (IL12A) also known as NKSF1, CLMF1 and P35, IL12A shows significant sequence similarity to IL-6, G-CSF, and exerts biological activities only when the IL12B is co-expressed. IL12B deficient mice are resistant to the induction of experimental chronic inflammatory diseases whereas IL12A knock-out mice develop more severe forms, suggesting opposite functions of the two subunits in the outcome of chronic inflammatory diseases.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 57-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
49.2 kDa | |
Human IL-12 A, Fc Tag (NK1-H5252) is expressed from human 293 cells (HEK293). It contains AA Arg 23 - Ser 219 (Accession # P29459). | |
-20°C | |
IL-12 A | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Cytokines and Growth Factors |
50 μg | |
Arg 23 - Ser 219 | |
IL12A,CLMF,IL-12 A,NFSK,NKSF1,P35 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |