Learn More
ACROBiosystems Human OX40 / TNFRSF4 / CD134 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marke: ACROBiosystems OX0-H5252-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 47.1 kDa. The protein migrates as 58-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP 003318.1 | |
100 μg | |
Leu 29 - Ala 216 | |
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
47.1 kDa | |
Human O x 40, Mouse IgG2a Fc Tag, low endotoxin (O x 0-H5252) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # P43489-1). | |
-20°C | |
OX40 | |
Human | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |