Learn More
ACROBiosystems Biotinylated Human CD200 / OX-2 Protein, Fc, Avitag™
Biotinylated Protein
Marke: ACROBiosystems OX2-H82F1-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
CD200 is also known as OX-2 membrane glycoprotein (OX-2), is a type-1 membrane glycoprotein, which contains two immunoglobulin domains (1 Ig-like C2-type (immunoglobulin-like) domain and Ig-like V-type (immunoglobulin-like) domain), and thus belongs to the immunoglobulin superfamily. CD200 / OX-2 is widely expressed in multiple cell types. CD200 interacts with a structurally related receptor (CD200R) expressed mainly on myeloid cells and is involved in regulation of macrophage and mast cell function. OX-2 / CD200 and CD200R associate via their respective N-terminal Ig-like domains. CD200 also plays an important role in prevention of graft rejection, autoimmune diseases and spontaneous abortion.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.4 kDa. The protein migrates as 65-76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
XP_005247539.1 | |
25 μg | |
Gln 31 - Gly 232 | |
CD200,MOX1,MOX2,MRC,OX-2,My033 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
51.4 kDa | |
Biotinylated Human CD200, Fc,Avitag (O x 2-H82F1) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # P41217-1). | |
-20°C | |
CD200 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |