Learn More
ACROBiosystems Human Fc gamma RIIA / CD32a (R167) Protein, His Tag (BLI verified)
BLI verified Protein
Marke: ACROBiosystems CDA-H5221-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Receptors for the Fc region of IgG (Fc γ R) are members of the Ig superfamily that function in the activation or inhibition of immune responses. Three classes of human Fc γ Rs: RI (CD64), RII (CD32), and RIII (CD16), which generate multiple isoforms, are recognized. There are three genes for human Fcγ RII /CD32 (A, B, and C) and one for mouse Fcγ RII B (CD32B). CD32 is a low affinity receptor for IgG. The activating isoform, CD32A, is expressed on monocytes, neutrophils, platelets and dendritic cells. CD32A is expressed on many immune cell types (macrophage, neutrophil, eosinophils, platelets, dendritic cells and Langerhan cells), where inhibitory ITIMbearing receptors may also be coexpressed and coengaged by specific ligands. CD32A delivers an activating signal upon ligand binding, and results in the initiation of inflammatory responses including cytolysis, phagocytosis, degranulation and cytokine production. The responses can be modulated by signals from the coexpressed inhibitory receptors such as CD32B, and the strength of the signal is dependent on the ratio of expression of the activating and inhibitory receptors.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.6 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH19931.1 | |
100 μg | |
Ala 36 - Ile 218 | |
CD32a,FCGR2A,CD32,FCG2 ,FCGR2A1,IGFR2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
21.6 kDa | |
Human CD32a (R167), His Tag (BLI verified) (CDA-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1 (R167)). | |
-20°C | |
Fc gamma RIIA / CD32a | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Fc Receptors |