Learn More
ACROBiosystems Human IL-1 RII / CD121b Protein, His Tag
Marke: ACROBiosystems IL2-H4226-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Interleukin-1 receptor type 2 (IL1R2) is also known as CD121 antigen-like family member B (CDw121b), IL-1 type II receptor, Interleukin-1 receptor type II, belongs to the interleukin-1 receptor family. Two distinct types of IL1 receptors which are able to bind IL1 specifically have been identified, designated as IL1RI (IL1RA) and IL1RII (IL1RB). IL1R2 is non-signaling receptor for IL1A, IL1B and IL1RN, reduces IL1B activities. Serves as a decoy receptor by competetive binding to IL1B and preventing its binding to IL1R1. IL1R2 modulates cellular response through non-signaling association with IL1RAP after binding to IL1B. IL1R2 (membrane and secreted forms) preferentially binds IL1B and poorly IL1A and IL1RN. The secreted IL1R2 recruits secreted IL1RAP with high affinity; this complex formation may be the dominant mechanism for neutralization of IL1B by secreted/soluble receptors.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.9 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_004624.1 | |
100 μg | |
Phe 14 - Glu 343 | |
CD121b,IL1RB,IL1R2,CDw121b | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |
38.9 kDa | |
Human IL-1 RII, His Tag (IL2-H4226) is expressed from human 293 cells (HEK293). It contains AA Phe 14 - Glu 343 (Accession # NP_004624.1). | |
-20°C | |
IL-1 RII | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |