Learn More
ACROBiosystems Human TWEAK R / TNFRSF12A Protein, Fc Tag
Marke: ACROBiosystems TNA-H526A-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 12A (TNFRSF12A) is also known as Fibroblast growth factor-inducible immediate-early response protein 14 (FN14 or FGF-inducible 14), Tweak-receptor, CD antigen CD266. TNFRSF12A contains one TNFR-Cys repeat. TNFRSF12A is highly expressed in heart, placenta and kidney. TNFRSF12A is receptor for TNFSF12/TWEAK, and is weak inducer of apoptosis in some cell types. TNFRSF12A / FN14 promotes angiogenesis and the proliferation of endothelial cells.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 32.5 kDa. As a result of glycosylation, the protein migrates as 32-45 kDa under reducing (R) condition, and 55-80 kDa under non-reducing (NR) condition (SDS-PAGSpezifikation
AAH02718 | |
100 μg | |
Glu 28 - Trp 79 | |
TNFRSF12A,FGF-inducible 14,FN14,TweakR,CD266 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
32.5 kDa | |
Human TWEAK R, Fc Tag (TNA-H526a) is expressed from human 293 cells (HEK293). It contains AA Glu 28 - Trp 79 (Accession # AAH02718). | |
-20°C | |
TWEAK R | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |