Learn More
ACROBiosystems Biotinylated Human GPA33 Protein, His Tag, ultra sensitivity (primary amine labeling)
Marke: ACROBiosystems GP3-H8224-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Glycoprotein A33 (GPA33) is also known as Cell surface A33 antigen, is a single-pass type I membrane protein which is expressed in normal gastrointestinal epithelium and in 95% of colon cancers. GPA33 The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 1 Ig-like C2-type (immunoglobulin-like) domain and 1 Ig-like V-type (immunoglobulin-like) domain characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily, which contains. GPA33 may play a role in cell-cell recognition and signaling.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH74830 | |
200 μg | |
Ile 22 - Val 235 | |
GPA33,A33 | |
UltraLys | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
24.4 kDa | |
MABSol™ Biotinylated Human GPA33, His Tag, primary amine labeling (GP3-H8224) is expressed from human HEK293 cells. It contains AA Ile 22 - Val 235 (Accession # AAH74830). It is the biotinylated form of Human GPA33, His Tag (GP3-H5224). | |
-20°C | |
GPA33 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |