Learn More
ACROBiosystems Biotinylated Influenza A [A/Guinea fowl/Hong Kong/WF10/99(H9N2)] Hemagglutinin 1 (HA1) Protein, His Tag, Avitag™
Biotinylated Protein
Marke: ACROBiosystems HA1-V82E2-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Influenza, commonly known as “the flu”, is an infectious disease of birds and mammals caused by RNA viruses of the family Orthomyxoviridae, the influenza viruses. The virus is divided into three main types (Influenzavirus A, Influenzavirus B, and Influenzavirus C), which are distinguished by differences in two major internal proteins (hemagglutinin (HA) and neuraminidase (NA), which are the most important targets for the immune system. The type A viruses are the most virulent human pathogens among the three influenza types and cause the most severe disease. The serotypes that have been confirmed in humans, ordered by the number of known human pandemic deaths, are: H1N1, H2N2, H3N2, H5N1, H7N7, H1N2, H9N2, H7N2, H7N3, H10N7, H7N9. H7N9 is a serotype of the species Influenzavirus A (avian influenza virus or bird flu virus). H7 normally circulates amongst avian populations with some variants known to occasionally infect humans. An H7N9 virus was first reported to have infected humans in 2013 in China.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 39.3 kDa. The protein migrates as 47-70 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAO46082.1 | |
25 µg | |
Asp 19 - Arg 338 | |
HA1,Hemagglutinin 1 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
39.3 kDa | |
Biotinylated Influenza A [A/Guinea fowl/Hong Kong/WF10/99(H9N2)] HA1, His Tag,Avitag (HA1-V82E2) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Arg 338 (Accession # Q80KD9-1). | |
-20°C | |
Hemagglutinin 1 (HA1) | |
Influenza | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |