Learn More
ACROBiosystems Mouse gp130 / CD130 / IL-6 R beta Protein, His Tag
Marke: ACROBiosystems ILT-M52H1-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Interleukin-6 receptor subunit beta (IL6ST) is also known as IL-6 receptor subunit beta, IL-6R subunit beta, IL-6R-beta, IL-6RB, Interleukin-6 signal transducer, Membrane glycoprotein 130 (gp130), CD130, Oncostatin-M receptor subunit alpha and Il6st,which is single-pass type I membrane protein. IL6ST /gp130 /CD130 can be found in tissues such as brain, heart, thymus, spleen, kidney, lung and liver and found in all the cell lines tested except BaF-B03.The expression of IL-6ST /gp130 is not restricted to IL6-responsive cells. The receptor systems for IL6, LIF, OSM, CNTF, IL11, CTF1 and BSF3 can utilize gp130 for initiating signal transmission. IL6ST /CD130 can bind to IL6 /IL6R (alpha chain) complex, resulting in the formation of high-affinity IL6 binding sites, and transduce the signal. IL6ST /GP130 does not bind IL6 and may have a role in embryonic development.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 68.5 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_034690 | |
100 μg | |
Gln 23 - Glu 617 | |
IL6ST,gp130,CD130,IL-6 RB,IL-6 R-beta,CDw130 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
68.5 kDa | |
Mouse gp130, His Tag (ILT-M52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Glu 617 (Accession # NP_034690). | |
-20°C | |
gp130 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |