Learn More
ACROBiosystems Human Transthyretin / Prealbumin Protein, His Tag
Marke: ACROBiosystems TTR-H5223-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Transthyretin (TTR) is also known as Prealbumin, ATTR, TBPA, PALB, which belongs to the transthyretin family. Transthyretin / TTR is a serum and cerebrospinal fluid carrier of the thyroid hormone thyroxine (T4) and retinol-binding protein bound to retinol. In cerebrospinal fluid TTR is the primary carrier of T4. TTR also acts as a carrier of retinol (vitamin A) through its association with retinol-binding protein (RBP) in the blood and the CSF. Less than 1% of TTR's T4 binding sites are occupied in blood. TTR misfolding and aggregation is known to be associated with the amyloid diseases.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 17 kDa and 33 kDa under reducing (R) condition (SDS-PAGE) due to the monomer and dimer respectively.Spezifikation
NP_000362 | |
100 μg | |
Gly 21 - Glu 147 | |
Transthyretin,TTR,Prealbumin,TBPA,ATTR,PALB,CTS,CTS1,HsT2651 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
14.9 kDa | |
Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession # NP_000362). | |
-20°C | |
Transthyretin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |